Learn More
Leap PAL PartsSupplier Diversity Partner Kit Pack Screw Clear 3.0mL 20x46mm Flat, 12 Pack  DFS Item
Manufacturer:  Leap PAL PartsSupplier Diversity Partner XQS2226
Kit Pack. CD Vial, Heavy-walled, 3.0-mL, 20x46mm, Screw, Clear, 20-400, PTFE/Silicone, 12 Pack
Catalog No. 50-635-237