Learn More
Abnova™ KPNA1 293T Cell Transient Overexpression Lysate
Human overexpression lysate
Manufacturer:  Abnova™ H00003836T02
Catalog No. 89-950-165
Add to cart
Specifications
Specifications
Western Blot
293T
NM_002264
pCMV-KPNA1 full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Human
KPNA1 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 60.2kDa
Transient overexpression cell lysate was tested with anti-KPNA1 antibody (Mfr. No. H00003836-B01P) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)