Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KPNA6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA553867
Description
Immunogen sequence: RRREEEGIQL RKQKREQQLF KRRNVELINE EAAMFDSLLM DSYVSSTTGE SVITREMVEM LFSDDSDLQL ATTQKFR Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
Karyopherin, a cytosolic and heterodimeric protein complex consisting of alpha and beta subunits, is responsible for targeting proteins with nuclear localization signals to the nuclear pore complex (NPC) by an energy requiring, Ran-dependent mechanism. The alpha subunit and imported substrate enter the nucleus and accumulate in the nucleoplasm, while the beta subunit accumulates at the NPC. KPNA6 belongs to a subfamily within the KPNA family that also includes IPOA5&6. Down-regulation of KPNA6 by RNAi expression in HeLa cells strongly inhibited cell proliferation, possibly due to blocking the nuclear import of specific factors essential for cell growth and proliferation. KPNA6 also interacts with the Rev protein of HIV-1 and promote its nuclear import.
Specifications
KPNA6 | |
Polyclonal | |
Unconjugated | |
KPNA6 | |
FLJ11249; importin alpha 7 subunit; importin alpha-S2; Importin subunit alpha-7; importin-alpha-S2; IPOA7; karyopherin (importin) alpha 5; karyopherin (importin) alpha 6; karyopherin alpha 6; karyopherin alpha 6 (importin alpha 7); karyopherin subunit alpha 6; karyopherin subunit alpha-6; Kpna5; Kpna6; KPNA7; MGC17918; NPI-2; RP4-622L5.1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23633 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O60684 | |
KPNA6 | |
Recombinant protein corresponding to Human KPNA6. Recombinant protein control fragment (Product #RP-91826). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction