Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KPNA6 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen KPNA6
Applications Immunohistochemistry, Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB093831
View Documents
Novus Biologicals
NBP304919100UL
100 μg Each for $320.71
Add to cart
 
NB093832
View Documents
Novus Biologicals
NBP30491920UL
20 μg Each for $138.37
Add to cart
 
Description

Description

KPNA6 Polyclonal antibody specifically detects KPNA6 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry
Specifications

Specifications

KPNA6
Unconjugated
Rabbit
Core ESC Like Genes, Stem Cell Markers
PBS with 50% glycerol, pH7.3.
23633
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Immunohistochemistry, Western Blot
Purified
RUO
Human, Mouse
importin alpha 7 subunit, importin subunit alpha-7, importin-alpha-S2, IPOA7FLJ11249, karyopherin alpha 6 (importin alpha 7), Karyopherin subunit alpha-6, KPNA7, MGC17918
Recombinant fusion protein containing a sequence corresponding to amino acids 277-536 of human KPNA6 (NP_036448.1).
Polyclonal
Affinity purified
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit