Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KPRP Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA546028
Description
Peptide sequence: PHRLDTEAPY CGPSSYNQGQ ESGAGCGPGD VFPERRGQDG HGDQGNAFAG Sequence homology: Human: 100%.
The function of this protein remains unknown.
Specifications
KPRP | |
Polyclonal | |
Unconjugated | |
KPRP | |
C1orf45; hKPRP; keratinocyte expressed, proline-rich protein; keratinocyte proline rich protein; keratinocyte proline-rich protein; KPRP | |
Rabbit | |
Affinity Chromatography | |
RUO | |
448834 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q5T749 | |
KPRP | |
Synthetic peptide directed towards the C-terminal of human KPRP (aa 520-569). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction