Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Krackeler Scientific Vial Racks

Racks for use with microvials

Manufacturer:  Krackeler Scientific 9293010722EA

Catalog No. HP93010722N


Add to cart

Description

Description

  • For use with 12mm, 2mL crimp-top microvials
  • Made of plastic

Specifications

Specifications

Blue
1 in.
Plastic
8 in.
2mL 12mm Crimp top microvials
2.5 cm
50
20.3 cm
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit