Krackeler Scientific Copper Tubing

Manufacturer: Krackeler Scientific 9250217107EA

 View more versions of this product

Catalog No. 50217107N

  Check Availability
Add to Cart

    Specifications

    Product Type Copper tubing
    Diameter (English) Outer 1/8 in.
    Diameter (Metric) Outer 3.17mm
    For Use With (Application) Ideal for gas supplies
    For Use With (Equipment) Gas cylinder regulators
    Material Copper
    Length (English) 12 ft.
    Length (Metric) 3.65m

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit