Learn More
Krackeler Scientific Crimp Caps
For headspace top vials
Manufacturer:  Krackeler Scientific 9251811211EA
 View more versions of this product
Catalog No. HP51811211N
Add to cart
Description
Description
Specifications
Specifications
PTFE/Silicone/PTFE
Aluminum
100 Pack
11 mm
Crimp Cap