Learn More
Krackeler Scientific Ferrules: Graphite/Vespel
SDP
$197.10 - $207.50
Specifications
Material Graphite, Vespel
Type Ferrule
View More Specs
Products 3
Catalog Number Mfr. No. Diameter (Metric) Inside For Use With (Equipment) Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (Metric) Inside For Use With (Equipment) Price Quantity & Availability  
50623512N
SDP
View Documents
Krackeler Scientific
9250623512EA
0.08 cm GC and GC/MS Capillary Column, Fits Column I.D. 0.045 to 0.053 cm
Each for $198.10
Add to Cart
51813323N
SDP
View Documents
Krackeler Scientific
9251813323EA
0.04 cm Fits Column I.D. 0.025 mm
Each for $207.50
Add to Cart
50623514N
SDP
View Documents
Krackeler Scientific
9250623514EA
0.05 cm GC and GC/MS Capillary Column, Fits Column I.D. 0.032 cm
Each for $197.10
Add to Cart
Description
Description
Specifications
Specifications
Graphite, Vespel
Ferrule