Krackeler Scientific Ferrules: Vespel

Manufacturer: Krackeler Scientific 9250623511EA

 View more versions of this product

Catalog No. 50623511N

  Check Availability
Add to Cart

    Specifications

    Column Type GC
    Material Vespel
    Diameter (English) Inner 0.031 in.
    Diameter (Metric) Inner 0.08cm
    For Use With (Application) GC
    For Use With (Equipment) Isothermal analysis use with GC and GC/MS Capillary Column, fits Column I.D. 0.045 to 0.053cm
    Product Type Ferrule

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit