Learn More
Krackeler Scientific LC and LC/MS Columns Amino Acid Separations Reagents
Provide excellent amino acid separation
Manufacturer:  Krackeler Scientific 9250613335EA
 View more versions of this product
Catalog No. 50613335N
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
LC and LC/MS column
OPA reagent
4°C
Borate Buffer o-phthalaldehyde (OPA) 0.4M 10mg/mL, 3-mercapto propionic Acid 10mg/mL
10 x 1mL