Learn More
Krackeler Scientific LC and LC/MS Columns Amino Acid Separations Reagents
Provide excellent amino acid separation
Manufacturer:  Krackeler Scientific 9250613337EA
 View more versions of this product
Catalog No. 50613337N
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
LC and LC/MS column
FMOC reagent
4°C
9-fluorenylmethylchloroformate 2.5mg/mL in Aceto nitrile
10 x 1mL