Learn More
Krackeler Scientific Screw Caps with Septa
For wide opening screw top vials
Manufacturer:  Krackeler Scientific 9251820722EA
 View more versions of this product
Catalog No. 51820722N
Add to cart
Description
Description
Specifications
Specifications
Red
With precision-fit integrated septa; Type: PTFE; Material: Silicone
Polypropylene
Polypropylene
PTFE/White Silicone