Learn More
Krackeler Scientific Screw Caps with Septa
For wide opening screw top vials
Manufacturer:  Krackeler Scientific 9251820724EA
 View more versions of this product
Catalog No. 51820724N
Add to cart
Description
Description
Specifications
Specifications
Green
With precision-fit integrated septa; Type: PTFE; Material: Silicone/PTFE
Polypropylene
Polypropylene
PTFE/Silicone/PTFE