Krackeler Scientific Snap Caps with 11mm Septa
For vials with wide opening inserts
Manufacturer: Krackeler Scientific 9251823457EA
 View more versions of this product
Catalog No. 51823457N
  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
    Specifications
    Color Green
    Material Polyethylene
    Vial Closure Size 11mm
    Type Snap Top Cap
    Material Closure Polyethylene
    Material (Liner) PTFE/Silicone
  • Description & Specifications
Related Products
Product Content Feedback