Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Krackeler Scientific Snap Caps with 11mm Septa
For vials with wide opening inserts
$37.37 - $65.65
Catalog Number | Mfr. No. | Color | Liner Material | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Liner Material | Price | Quantity & Availability | ||||
51820541N
|
Krackeler Scientific
9251820541EA |
Blue | PTFE/Silicone |
Each for $65.65
|
|
51820550N
|
Krackeler Scientific
9251820550EA |
Clear | PTFE/Silicone Rubber |
Each for $42.17
|
|
51823457N
|
Krackeler Scientific
9251823457EA |
Green | PTFE/Silicone |
Each for $37.37
|
|
Specifications
11 mm | |
Polyethylene |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title