Krackeler Scientific Snap Caps with 11mm Septa
For vials with wide opening inserts
Manufacturer: Krackeler Scientific 9251820541EA
 View more versions of this product
Catalog No. 51820541N
  Check Availability
Add to Cart
    Specifications
    Material Polyethylene
    Type Snap Top Cap
    Material Closure Polyethylene
    Color Blue
    Vial Closure Size 11mm
    Material (Liner) PTFE/Silicone
  • Description & Specifications
Related Products
Product Content Feedback