Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Krackeler Scientific Wide Opening Crimp Top Glass Vials

Manufacturer:  Krackeler Scientific 9251813376EA

 View more versions of this product

Catalog No. 51813376N


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

Crimp
12mm
Silica Glass
2mL
Yes
0.5 dr.
2.0mL
Amber
Crimp Top Vial
32mm
Wide
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit