Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KRAS Mouse anti-Human, Clone: 2F8, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP25941350UL

 View more versions of this product

Catalog No. NBP25941350


Add to cart

Description

Description

KRAS Monoclonal antibody specifically detects KRAS in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

KRAS
2F8
Unconjugated
Liquid. In PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide
cellular c-Ki-ras2 proto-oncogene, c-Ki-ras, C-K-RAS, GTPase KRas, Ki-Ras, Kirsten rat sarcoma-2 viral (v-Ki-ras2) oncogene homolog, K-Ras 2, K-ras p21 protein, KRAS1, K-RAS2A, K-RAS2B, KRAS2NS3, K-RAS4A, K-RAS4B, NS, oncogene KRAS2, PR310 c-K-ras oncogene, RASK2c-Kirsten-ras protein, transforming protein p21, v-Ki-ras2 Kirsten rat sarcoma 2 viral oncogene homolog, v-Ki-ras2 Kirsten rat sarcoma viral oncogene homolog
Mouse
IgG2b Kappa
50 ul
Cell Cycle and Replication, Core ESC Like Genes, mTOR Pathway, Stem Cell Markers
Primary
3845
ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
1 mg/mL
Western Blot 1:1000, Flow Cytometry 1:10-1:1023, ELISA, Immunocytochemistry/Immunofluorescence 1:100
Purified
KRAS
Recombinant human KRAS (1-186aa) purified from E. coli
Protein A purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only