Learn More
KRas Rabbit anti-Human, Unconjugated, Clone: 2J13, MilliporeSigma™
Rabbit Monoclonal Antibody
Manufacturer:  MilliporeSigma ZRB00384525UL
 View more versions of this product
Catalog No. ZRB003825MI
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
KRas Monoclonal antibody specifically detects KRas in Human samples. It is validated for Immunohistochemistry (Paraffin), RNAi/siRNA/Gene Knockdown, Western Blotting.
Specifications
Specifications
KRas
2J13
Unconjugated
Purified
KRAS, KRAS2, RASK2
GST/His-tagged full-length recombinant human KRas.
∼21 kDa observed; 21.66 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
25 μL
Recommend storage of lyophilized product at 2 to 8°C. Avoid repeated freeze-thaws.
Monoclonal
Immunohistochemistry (Paraffin), Knockdown, Western Blot
120 μg/mL after reconstitution at 25 μL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.
NP_203524
GTPase KRas;K-Ras 2;Ki-Ras;c-K-ras;c-Ki-ras
Rabbit
IgG
Protein A Purified
Signaling
Primary
Human