Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KRas Rabbit anti-Human, Unconjugated, Clone: 2J13, MilliporeSigma™

Rabbit Monoclonal Antibody

Specifications

Antigen KRas
Applications Immunohistochemistry (Paraffin), Knockdown, Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
ZRB003825MI
View Documents
MilliporeSigma
ZRB00384525UL
25 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
ZRB003845MI
View Documents
MilliporeSigma
ZRB0038454X25UL
4 x 25 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

KRas Monoclonal antibody specifically detects KRas in Human samples. It is validated for Immunohistochemistry (Paraffin), RNAi/siRNA/Gene Knockdown, Western Blotting.
Specifications

Specifications

KRas
Unconjugated
Rabbit
GTPase KRas;K-Ras 2;Ki-Ras;c-K-ras;c-Ki-ras
GST/His-tagged full-length recombinant human KRas.
Primary
Recommend storage of lyophilized product at 2 to 8°C. Avoid repeated freeze-thaws.
Immunohistochemistry (Paraffin), Knockdown, Western Blot
Purified
Human
KRAS, KRAS2, RASK2
Monoclonal
Protein A Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit