Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Krebs-Ringer Solution, HEPES-buffered, Alfa Aesar™

Standard physiological buffer that is suitable for various calcium imaging experiments. It is also used as a preserving agent.

$72.00 - $118.00

Specifications

Chemical Name or Material Krebs-Ringer Solution
Packaging Plastic bottle
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAJ67795K2 View Documents Alfa Aesar
J67795K2
1L Each for $118.00
Order in Bulk

Request bulk or custom formats

Add to cart
 
AAJ67795AP View Documents Alfa Aesar
J67795AP
500mL Each for $72.00
Order in Bulk

Request bulk or custom formats

Add to cart
 
Specifications

Specifications

Krebs-Ringer Solution
Plastic bottle
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Ambient temperatures

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit