Kresto™ ATP Cupran™ Special Heavy-Duty Skin Cleaner

High-performance skin cleaner for oil-based paints, stains, urethane sealants, adhesives, tar, and asphalt

Manufacturer: Stockhausen Inc. 9 81874 06

 View more versions of this product

Catalog No. 19-819-814

  Check Availability
Add to Cart

    Specifications

    Product Type Cupran™ Special Skin Cleaner
    Size (English) 67.62 oz.
    Size (Metric) 2000mL
    Quantity 6 Case
    Composition Astopon walnut shell scrubbers
    For Use With (Application) Removes oil based paints, stains, urethane sealants, adhesives, tar, and asphalt from hands

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit