Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KRIT1 Polyclonal Antibody
KRIT1 Polyclonal Antibody
Supplier: Thermo Scientific PA595383
Description
KRIT1 Polyclonal Antibody for Western Blot

Specifications
KRIT1 | |
Polyclonal | |
Unconjugated | |
KRIT1 | |
2010007K12Rik; A630036P20Rik; AA432855; AI450393; AI643869; ankyrin repeat-containing protein Krit1; BB155247; BB235701; CAM; Ccm1; Cerebral cavernous malformations 1 protein; cerebral cavernous malformations 1 protein homolog; Krev interaction trapped 1; krev interaction trapped protein 1; KRIT1; KRIT1, ankyrin repeat containing; RGD1305929 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
362317, 79264, 889 | |
-20°C | |
Lyophilized |
Western Blot | |
500 μg/mL | |
PBS with 5mg BSA and 0.05mg sodium azide | |
O00522, Q6S5J6 | |
KRIT1 | |
A synthetic peptide corresponding to a sequence at the C-terminus of human KRIT1 (703-736aa ENKMSFIVHTKQAGLVVKLLMKLNGQLMPTERNS). | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction