Learn More
MilliporeSigma™ Supelco™ Kromasil™ AmyCoat™ Chiral Reversed Phase HPLC Column, 3 µm Particle Size
Polysaccharide-based Kromasil™ AmyCoat™ and CelluCoat™ stretch the limits for chiral chromatography. The silica is based on a proprietary matrix and coated with a functionalized amylose or cellulose selector.
Manufacturer:  MilliporeSigma™ Supelco™ K08670500
 View more versions of this product
Catalog No. 11-100-2919
Add to cart
Description
Description
Kromasil™ AmyCoat™ and CelluCoat™ give high resolution, excellent selectivity and stable performance when switching between compatible mobile phases. Users do not have to worry about pressure limits, as both Kromasil™ AmyCoat™ and CelluCoat™ can withstand flow rates equivalent to pressures of up to 400 bar – i.e. the limit for most standard HPLC systems.
Specifications
Specifications
Carbamoyl Phase
4.6 mm
3 μm
Chiral HPLC
Kromasil AmyCoat
Chiral
50 mm
>1000 Å
Carbamoyl
SDS
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
Kromasil and AmyCoat are registered trademarks of EKA Chemicals AB