Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Supelco™ Kromasil C18 HPLC Column, 300 Å, 16 μm Particle Size
Protein and biomolecule separations from analytical to process scale.
Manufacturer: MilliporeSigma™ Supelco™ K08971058
Description
The Kromasil Classic 300 Å family of products is designed to be the perfect choice for proteins and biomolecules larger than 8–10 kDa. This 300 Å material has a narrow pore size distribution that ensures good mass transfer for larger molecules, resulting in narrow peaks and no size-exclusion effects.Specifications
10 mm | |
C18 (octadecyl) Phase | |
Reversed Phase | |
Kromasil | |
HPLC |
250 mm | |
16 μm | |
300 Å | |
C18 (octadecyl) |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Kromasil is a registered trademark of EKA Chemicals AB