Learn More
MilliporeSigma Supelco Kromasil C18 HPLC Column, 300 Å, 16 µm Particle Size
Protein and biomolecule separations from analytical to process scale.
$541.50 - $14190.00
Catalog Number | Mfr. No. | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | ||||
11-100-3571
|
MilliporeSigma Supelco
K08971040 |
Each for $541.50
|
|
11-100-3445
|
MilliporeSigma Supelco
K08971049 |
Each for $601.00
|
|
11-100-2987
|
MilliporeSigma Supelco
K08971058 |
Each for $1,512.00
|
|
11-100-2847
|
MilliporeSigma Supelco
K08971067 |
Each for $3,250.00
|
|
11-100-2740
|
MilliporeSigma Supelco
K08971076 |
Each for $14,190.00
|
|
Description
The Kromasil Classic 300 Å family of products is designed to be the perfect choice for proteins and biomolecules larger than 8–10 kDa. This 300 Å material has a narrow pore size distribution that ensures good mass transfer for larger molecules, resulting in narrow peaks and no size-exclusion effects.Kromasil is a registered trademark of EKA Chemicals AB