Learn More
MilliporeSigma™ Supelco™ Kromasil C8 HPLC Column, 300 Å, 5 µm Particle Size
Protein and biomolecule separations from analytical to process scale.
Manufacturer:  MilliporeSigma™ Supelco™ K08670771
 View more versions of this product
Catalog No. 11-100-3381
Add to cart
Description
Description
The Kromasil Classic 300 Å family of products is designed to be the perfect choice for proteins and biomolecules larger than 8–10 kDa. This 300 Å material has a narrow pore size distribution that ensures good mass transfer for larger molecules, resulting in narrow peaks and no size-exclusion effects.
Specifications
Specifications
Reversed Phase
3 mm
5 μm
HPLC
Kromasil
C8 (Octyl) Phase
50 mm
300 Å
C8 (Octyl)
SDS
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
Kromasil is a registered trademark of EKA Chemicals AB