Learn More
MilliporeSigma™ Supelco™ Kromasil C8 HPLC Column, 300 Å, 5 µm Particle Size
Protein and biomolecule separations from analytical to process scale.
$416.50 - $3860.00
Products 12
Catalog Number Mfr. No. Diameter (Metric) Length (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (Metric) Length (Metric) Price Quantity & Availability  
11-100-3382
View Documents
MilliporeSigma™ Supelco™
K08670775
Each for $610.00
Add to cart
11-100-3346
View Documents
MilliporeSigma™ Supelco™
K08670773
Each for $631.00
Add to cart
11-100-3295
View Documents
MilliporeSigma™ Supelco™
K08670774
Each for $662.50
Add to cart
11-100-3381
View Documents
MilliporeSigma™ Supelco™
K08670771
Each for $610.00
Add to cart
11-100-3345
View Documents
MilliporeSigma™ Supelco™
K08670769
Each for $631.00
Add to cart
11-100-3294
View Documents
MilliporeSigma™ Supelco™
K08670770
Each for $662.50
Add to cart
11-100-3727
View Documents
MilliporeSigma™ Supelco™
K08670780
Each for $416.50
Add to cart
11-100-3639
View Documents
MilliporeSigma™ Supelco™
K08670777
Each for $487.00
Add to cart
11-100-3542
View Documents
MilliporeSigma™ Supelco™
K08670778
Each for $541.50
Add to cart
11-100-3417
View Documents
MilliporeSigma™ Supelco™
K08670779
Each for $601.00
Add to cart
11-100-2967
View Documents
MilliporeSigma™ Supelco™
K08670772
Each for $1,512.00
Add to cart
11-100-2811
View Documents
MilliporeSigma™ Supelco™
K08670776
Each for $3,860.00
Add to cart
Description
Description
The Kromasil Classic 300 Å family of products is designed to be the perfect choice for proteins and biomolecules larger than 8–10 kDa. This 300 Å material has a narrow pore size distribution that ensures good mass transfer for larger molecules, resulting in narrow peaks and no size-exclusion effects.
SDS
Kromasil is a registered trademark of EKA Chemicals AB