Learn More
MilliporeSigma™ Supelco™ Kromasil Diol HPLC Column, 60 Å, 5 µm Particle Size
For separation of small molecules from analytical to process scale.
Manufacturer:  MilliporeSigma™ Supelco™ K08670841
 View more versions of this product
Catalog No. 11-100-3386
Add to cart
Description
Description
The Kromasil Classic 60 Å family of products is the choice for small, organic molecules when a large, accessible surface area is key for separating peaks in analysis. It also has the added properties of loadability and capacity required for purification.
Specifications
Specifications
Diol Phase
2.1 mm
5 μm
HPLC
Kromasil
Normal Phase and HILIC
50 mm
60 Å
Diol
SDS
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
Kromasil is a registered trademark of EKA Chemicals AB