Learn More
MilliporeSigma™ Supelco™ Kromasil™ Eternity™ C18 HPLC Column, 100 Å, 5 µm Particle Size
Kromasil™ Eternity™ and Eternity™XT can be used in a wider pH window than regular silica-based HPLC materials, giving more flexibility than ever when running analytical applications or optimizing for large-scale applications.
Manufacturer:  MilliporeSigma™ Supelco™ K08670901
 View more versions of this product
Catalog No. 11-100-3220
Add to cart
Description
Description
For regular silica-based stationary phases, exposure to extreme pH (especially basic) will have a negative impact on the chemical stability and therefore column lifetime. However, the silica/organosilane surface of the Kromasil™ Eternity™ platform offers a chemical stability that will secure a long-lasting stationary phase, even under tough pH conditions and higher temperatures.
Specifications
Specifications
C18 (octadecyl) Phase
4.6 mm
5 μm
330 m2/g
C18 (octadecyl)
Reversed Phase
100 mm
100 Å
HPLC
Kromasil Eternity
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
Kromasil and Eternity are trademarks of EKA Chemicals AB