Learn More
MilliporeSigma™ Supelco™ Kromasil Silica HPLC Column, 100 Å, 3.5 µm Particle Size
For small molecules and peptides.
$505.00 - $716.50
Specifications
Particle Size 3.5 μm
Phase Normal Phase and HILIC
Pore Size 100 Å
Product Line Kromasil
Matrix Silica Phase
View More Specs
Products 5
Catalog Number Mfr. No. Diameter (Metric) Length (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (Metric) Length (Metric) Price Quantity & Availability  
11-100-3250
View Documents
MilliporeSigma™ Supelco™
K08670437
2.1 mm 100 mm
Each for $684.50
Add to cart
11-100-3223
View Documents
MilliporeSigma™ Supelco™
K08670438
2.1 mm 150 mm
Each for $716.50
Add to cart
11-100-3603
View Documents
MilliporeSigma™ Supelco™
K08670441
4.6 mm 50 mm
Each for $505.00
Add to cart
11-100-3466
View Documents
MilliporeSigma™ Supelco™
K08670439
4.6 mm 100 mm
Each for $593.00
Add to cart
11-100-3308
View Documents
MilliporeSigma™ Supelco™
K08670440
4.6 mm 150 mm
Each for $656.50
Add to cart
Description
Description
The Kromasil Classic 100 Å family of products is used to separate and purify molecules of up to about 10,000 Da.
Specifications
Specifications
3.5 μm
100 Å
Silica Phase
Silica
Normal Phase and HILIC
Kromasil
HPLC
SDS
Kromasil is a registered trademark of EKA Chemicals AB