Learn More
MilliporeSigma™ Supelco™ Kromasil Silica HPLC Column, 100 Å, 3.5 µm Particle Size
For small molecules and peptides.
Manufacturer:  MilliporeSigma™ Supelco™ K08670440
 View more versions of this product
Catalog No. 11-100-3308
Add to cart
Description
Description
The Kromasil Classic 100 Å family of products is used to separate and purify molecules of up to about 10,000 Da.
Specifications
Specifications
Silica Phase
4.6 mm
3.5 μm
HPLC
Kromasil
Normal Phase and HILIC
150 mm
100 Å
Silica
SDS
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
Kromasil is a registered trademark of EKA Chemicals AB