Learn More
MilliporeSigma™ Supelco™ Kromasil Silica HPLC Column, 60 Å, 13 µm Particle Size
For separation of small molecules from analytical to process scale.
Manufacturer:  MilliporeSigma™ Supelco™ K08971005
 View more versions of this product
Catalog No. 11-100-3432
Add to cart
Description
Description
The Kromasil Classic 60 Å family of products is the choice for small, organic molecules when a large, accessible surface area is key for separating peaks in analysis. It also has the added properties of loadability and capacity required for purification.
Specifications
Specifications
Normal Phase and HILIC
4.6 mm
13 μm
HPLC
Kromasil
Silica Phase
250 mm
60 Å
Silica
SDS
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
Kromasil is a registered trademark of EKA Chemicals AB