Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KRR1 Polyclonal Antibody

KRR1 Polyclonal Antibody

Supplier:  Thermo Scientific PA521693

Catalog No. PIPA521693


Only null left
Add to Cart

Description

Description

KRR1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

TRUSTED_SUSTAINABILITY
Specifications

Specifications

KRR1
Polyclonal
Unconjugated
KRR1
2610511F02Rik; AI255219; AI428520; D10Ertd773e; HIV-1 Rev binding protein 2; HIV-1 Rev-binding protein 2; HIV-1 Rev-binding protein 2 homolog; Hrb2; Krr1; KRR1 small subunit processome component homolog; KRR1, small subunit (SSU) processome component, homolog; KRR1, small subunit (SSU) processome component, homolog (yeast); KRR1, small subunit processome component; KRR1, small subunit processome component homolog; KRR-R motif-containing protein 1; Rev interacting protein; rev-interacting protein 1; Rip-1
Rabbit
Antigen affinity chromatography
RUO
11103
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
1 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Q13601
KRR1
Recombinant fragment corresponding to a region within amino acids 47 and 344 of KRR1 (Uniprot ID#Q13601).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.