Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KRT10/KRT13 Mouse anti-Human, Clone: DE-K13, Abnova™

Mouse Monoclonal Antibody

Manufacturer:  Abnova Corporation MAB14966

Catalog No. 89-034-986


Add to cart

Description

Description

Mouse monoclonal antibody raised against native human KRT10/KRT13.

  • High specificity
  • High reactivity
Specifications

Specifications

KRT10/KRT13
DE-K13
Mouse monoclonal antibody raised against native human KRT10/KRT13.
In 10 mM PBS (0.05% BSA, 0.05% sodium azide).
Liquid
KRT10
Cytoskeletal preparation extracted from human ectocervical epithelium.
Protein A/G purification
RUO
Primary
3858|3860
Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
Flow Cytometry (0.5-1 ug/106 cells in 0.1 mL) Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) Western Blotting (0.25-0.5 ug/mL) The optimal working dilution should
P13645;P13646
CK10|K10|KPP
Mouse
IgG2a, kappa
100 ug
Store at 4°C.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only (RUO)