Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KRT10 Mouse anti-Human, Clone: DE-K10, Abnova™

Mouse monoclonal antibody raised against human KRT10.

Manufacturer:  Abnova Corporation MAB12724

Catalog No. 89-934-418

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This gene encodes a member of the type I (acidic) cytokeratin family, which belongs to the superfamily of intermediate filament (IF) proteins. Keratins are heteropolymeric structural proteins which form the intermediate filament. These filaments, along with actin microfilaments and microtubules, compose the cytoskeleton of epithelial cells. Mutations in this gene are associated with epidermolytic hyperkeratosis. This gene is located within a cluster of keratin family members on chromosome 17q21. [provided by RefSeq]

Specifications

Specifications

KRT10
DE-K10
Mouse monoclonal antibody raised against human KRT10.
In 0.15M PBS (1% BSA, 0.02% sodium azide).
CK10/K10/KPP
Mouse
100 uL
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
Immunohistochemistry (Frozen), Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
Immunohistochemistry (Frozen sections) (1:5-1:10) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:100) Western Blot The optimal working dilution should be determined by the end user.
Liquid
KRT10
Cytoskeletal preparation from human epidermis.
RUO
Primary
3858
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit