Learn More
KRT10 Mouse anti-Human, Unconjugated, Clone: rKRT10/1275, Abnova™
Mouse Monoclonal Antibody.
Manufacturer:  Abnova Corporation RAB00506
Catalog No. 89-337-931
Add to cart
Description
Description
KRT10 Monoclonal antibody specifically detects KRT10 in Human samples. It is validated for IHC-P.
Specifications
Specifications
KRT10
rKRT10/1275
keratin 10
Liquid
Mouse
IgG1, Kappa
Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
Immunohistochemistry (Paraffin)
Unconjugated
KRT10
CK10, K10, KPP
Original antibody is raised against recombinant protein corresponding to full length human KRT10
100 μg
Primary
3858