Learn More
KRT10 Rabbit anti-Human, Unconjugated, Clone: KRT10/1990R, Abnova™
Rabbit Monoclonal Antibody.
Manufacturer:  Abnova Corporation RAB00509
Catalog No. 89-337-934
Add to cart
Description
Description
KRT10 Monoclonal antibody specifically detects KRT10 in Human samples. It is validated for IHC-P.
Specifications
Specifications
KRT10
KRT10/1990R
keratin 10
Liquid
Rabbit
IgG
Store at 4°C
Monoclonal
Human
Immunohistochemistry (Paraffin)
Unconjugated
KRT10
CK10, K10, KPP
Original antibody is raised against recombinant protein corresponding to full length human KRT10
100 μg
Primary
3858