Learn More
KRT15 Mouse anti-Human, Clone: 15K01, Abnova™
Mouse monoclonal antibody raised against human KRT15.
Manufacturer:  Abnova Corporation MAB12547
Catalog No. 89-934-423
Add to cart
Description
Description
The protein encoded by this gene is a member of the keratin gene family. The keratins are intermediate filament proteins responsible for the structural integrity of epithelial cells and are subdivided into cytokeratins and hair keratins. Most of the type I cytokeratins consist of acidic proteins which are arranged in pairs of heterotypic keratin chains and are clustered in a region on chromosome 17q21.2. [provided by RefSeq]
Specifications
Specifications
KRT15
15K01
Mouse monoclonal antibody raised against human KRT15.
In PBS, pH 7.4 (0.2% BSA, 0.09% sodium azide) and before the addition of glycerol to 40%.
CK15/K15/K1CO
Mouse
IgG2a, kappa
50 uL
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
Immunofluorescence, Immunohistochemistry (PFA fixed)
Unconjugated
Immunofluorescence Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (5 ug/mL) The optimal working dilution should be determined by the end user.
Liquid
KRT15
A synthetic peptide corresponding to 17 residues at C-terminus region of human KRT15.
Protein A purification
RUO
Primary
3866