Learn More
KRT18 Mouse anti-Human, FITC, Clone: Ks 18.04, Abnova™
Mouse monoclonal antibody raised against human KRT18.
Manufacturer:  Abnova Corporation MAB12582
Catalog No. 89-934-425
Add to cart
Description
Description
KRT18 encodes the type I intermediate filament chain keratin 18. Keratin 18, together with its filament partner keratin 8, are perhaps the most commonly found members of the intermediate filament gene family. They are expressed in single layer epithelial tissues of the body. Mutations in this gene have been linked to cryptogenic cirrhosis. Two transcript variants encoding the same protein have been found for this gene. [provided by RefSeq]
Specifications
Specifications
KRT18
Ks 18.04
Mouse monoclonal antibody raised against human KRT18.
In PBS (0.09% sodium azide).
CYK18/K18
Mouse
IgG1
50 uL
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (PFA fixed)
FITC
Flow Cytometry Immunohistochemistry (Frozen sections) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:10) The optimal working dilution should be determined by the end user.
Liquid
KRT18
Human KRT18 from HeLa cytoskeletal preparation.
Protein A purification
RUO
Primary
3875