Learn More
KRT19 293T Cell Overexpression Lysate 2 (Denatured), Abnova
Manufacturer: Abnova Corporation H00003880T02
Description
Western Blotting
Specifications
Western Blot | |
KRT19 293T Cell Transient Overexpression Lysate(Denatured) | |
44.11 | |
pCMV-KRT19 full-length | |
100 μL | |
Human |
293T | |
BC007628.1 | |
6.8 | |
Transient overexpression cell Lysate was tested with Anti-KRT19 antibody (H00003880-B02) by western blotting | |
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue) |