Learn More
Aviva Systems Biology Corp KRT19 Antibody - N-terminal region (ARP47464_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP47464_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KRT19. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human KRT19. Alternate names: CK19, K19, K1CS, MGC15366ApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-59556