Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KRT19 Rabbit anti-Human, Mouse, Unconjugated, Clone: KRT19/1959R, Abnova™

Rabbit Monoclonal Antibody.

Manufacturer:  Abnova Corporation RAB00571

Catalog No. 89-339-527


Add to cart

Description

Description

KRT19 Monoclonal antibody specifically detects KRT19 in Human, Mouse samples. It is validated for WB-Ce, IHC-P.
Specifications

Specifications

KRT19
KRT19/1959R
keratin 19
Liquid
Rabbit
IgG
Store at -20 to -80°C
Monoclonal
Human, Mouse
Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
KRT19
CK19, K19, K1CS, MGC15366
Recombinant protein corresponding to full-length human KRT19
100 μg
Primary
3880
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit