Learn More
KRT20 293T Cell Overexpression Lysate (Denatured), Abnova
Manufacturer:  Abnova Corporation H00054474T01
Catalog No. 89-002-378
Add to cart
Description
Description
Western Blotting
Specifications
Specifications
Western Blot
293T
NM_019010.1
6.8
Transient overexpression cell Lysate was tested with Anti-KRT20 antibody (H00054474-B01) by western blotting
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Human
KRT20 293T Cell Transient Overexpression Lysate(Denatured)
48.5
pCMV-KRT20 full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.