Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KRT25 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548148

Catalog No. PA548148

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Essential for the proper assembly of type I and type II keratin protein complexes and formation of keratin intermediate filaments in the inner root sheath (irs).
Specifications

Specifications

KRT25
Polyclonal
PBS with 0.09% sodium azide
Q7Z3Z0
Keratin, type I cytoskeletal 25, Cytokeratin-25, CK-25, Keratin-25, K25, Keratin-25A, K25A, Type I inner root sheath-specific keratin-K25irs1, KRT25 (HGNC:30839)
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
147183
Flow Cytometry, Western Blot
Unconjugated
KRT25
Liquid
KRT25
KLH conjugated synthetic peptide between 380-408 amino acids from the C-terminal region of human KRT25.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit