Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KRT28 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA566878
Description
Immunogen sequence: FCSLGTLQFCI DKVNISQNTM SLQFSNGSRH VCLRSGAGSV RPLNGGAGFA GSSACGGSVA GSEFSCALGG GLGSVPGGSH AGGALGNAAC IGFAGSEG Highest antigen sequence identity to the following orthologs - mouse 57%, rat 60%.
Essential for the proper assembly of types I and II keratin protein complexes and the formation of keratin intermediate filaments in the inner root sheath (irs).
Specifications
KRT28 | |
Polyclonal | |
Unconjugated | |
KRT28 | |
CK-28; cytokeratin-28; K25D; K25IRS4; K28; keratin 25D; keratin 28; keratin 28, type I; keratin, type I cytoskeletal 28; Keratin-25D; keratin-28; KRT25D; KRT28; KRT28 (HGNC:30842); type I inner root sheath specific keratin 25 irs4; type I inner root sheath-specific keratin-K25irs4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
162605 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q7Z3Y7 | |
KRT28 | |
Recombinant Human KRT28. Recombinant protein control fragment (Product #RP-107527). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction