Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KRT28 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA548737
Description
Essential for the proper assembly of types I and II keratin protein complexes and the formation of keratin intermediate filaments in the inner root sheath (irs).
Specifications
KRT28 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q7Z3Y7 | |
KRT28 | |
KLH conjugated synthetic peptide between 432-460 amino acids from the C-terminal region of human KRT28. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
KRT28 | |
Keratin, type I cytoskeletal 28, Cytokeratin-28, CK-28, Keratin-25D, K25D, Keratin-28, K28, Type I inner root sheath-specific keratin-K25irs4, KRT28 (HGNC:30842) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
162605 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction