Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KRT5 Mouse anti-Human, Monkey, Mouse, Clone: 2C2, Abnova™

Mouse monoclonal antibody raised against partial recombinant human KRT5.

Manufacturer:  Abnova Corporation MAB12460

Catalog No. 89-934-434

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

The protein encoded by this gene is a member of the keratin gene family. The type II cytokeratins consist of basic or neutral proteins which are arranged in pairs of heterotypic keratin chains coexpressed during differentiation of simple and stratified epithelial tissues. This type II cytokeratin is specifically expressed in the basal layer of the epidermis with family member KRT14. Mutations in these genes have been associated with a complex of diseases termed epidermolysis bullosa simplex. The type II cytokeratins are clustered in a region of chromosome 12q12-q13. [provided by RefSeq]

Specifications

Specifications

KRT5
2C2
Mouse monoclonal antibody raised against partial recombinant human KRT5.
In PBS (0.05% protein stabilizer, 0.05% sodium azide)
CK5/DDD/EBS2/K5/KRT5A
Mouse
IgG1
50 ug
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human, Monkey, Murine
ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
ELISA (1:10000) Flow Cytometry (1:200-1:400) Immunocytochemistry (1:200-1:1000) Immunofluorescence (1:1000) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) Western Blot (1:500-1:2000) The optimal working d
Liquid
KRT5
Recombinant protein corresponding to amino acids 158-272 of human KRT5.
Protein G purification
RUO
Primary
3852
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit