Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KRT6 Mouse anti-Human, Clone: KRT6/1702, Abnova™

Mouse Monoclonal Antibody

Manufacturer:  Abnova Corporation MAB14574

Catalog No. 89-034-657


Add to cart

Description

Description

Mouse monoclonal antibody raised against full length recombinant human KRT6.

  • High specificity
  • High reactivity
Specifications

Specifications

KRT6
KRT6/1702
Mouse monoclonal antibody raised against full length recombinant human KRT6.
In 10 mM PBS (0.05% BSA, 0.05% sodium azide).
Liquid
KRT6A
Recombinant protein corresponding to full length human KRT6.
Protein A/G purification
RUO
Primary
3853|3854|286887
Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed)
Unconjugated
Flow Cytometry (0.5-1 ug/106 cells) Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.1-0.2 ug/mL) The optimal working dilution should be determined by the end user.
P02538;P04259;P48668
CK6A|CK6C|CK6D|K6A|K6C|K6D|KRT6C|KRT6D
Mouse
IgG2a, kappa
100 ug
Store at 4°C.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only (RUO)